Tisk…

Anti-SMARCA2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB15597-100EA 23870 CZK
ABCAAB15597-100
Anti-SMARCA2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SMARCA2 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR