Tisk…

Anti-SERPINA3 Mouse monoclonal antibody [clone: ACT14C7]

ABCAAB15606-200EA 17160 CZK
ABCAAB15606-200
Anti-SERPINA3 Mouse monoclonal antibody [clone: ACT14C7]
Type: Primary
Antigen: SERPINA3 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3)
Clonality: monoclonal
Clone: ACT14C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR