Tisk…
Anti-Ly6c Rat Monoclonal Antibody (Biotin) [clone: ER-MP20]
Dodavatel: Abcam
ABCAAB15674-50EA 25740 CZK
ABCAAB15674-50
Anti-Ly6c Rat Monoclonal Antibody (Biotin) [clone: ER-MP20]
Protilátky
Type: Primary
Antigen: Ly6c
Clonality: monoclonal
Clone: ER-MP20
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse