Tisk…

Anti-RGS19 Chicken polyclonal antibody

ABCAAB15881-50EA 18270 CZK
ABCAAB15881-50
Anti-RGS19 Chicken polyclonal antibody
Type: Primary
Antigen: RGS19 (regulator of G-protein signaling 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR