Tisk…

Anti-Staphylococcus Enterotoxin H Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Abcam
ABCAAB15909-250EA 21360 CZK
ABCAAB15909-250
Anti-Staphylococcus Enterotoxin H Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: Staphylococcus Enterotoxin H
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR