Tisk…

Anti-CPEB1 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB15917-100EA 21330 CZK
ABCAAB15917-100
Anti-CPEB1 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CPEB1 (cytoplasmic polyadenylation element binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR