Tisk…

Anti-Pecanex Goat polyclonal antibody

ABCAAB15939-100EA 10660 CZK
ABCAAB15939-100
Anti-Pecanex Goat polyclonal antibody
Type: Primary
Antigen: Pecanex
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR