Tisk…
Anti-SOCS3 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB16030-100EA 28870 CZK
ABCAAB16030-100
Anti-SOCS3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SOCS3 (suppressor of cytokine signaling 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat