Tisk…
Anti-FOXP2 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB16046-100EA 28800 CZK
ABCAAB16046-100
Anti-FOXP2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FOXP2 (forkhead box P2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat