Tisk…

Anti-ADAR Sheep Polyclonal Antibody

Dodavatel: Abcam
ABCAAB16138-50EA 22340 CZK
ABCAAB16138-50
Anti-ADAR Sheep Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ADAR (adenosine deaminase, RNA-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR