Tisk…

Anti-TNFRSF10B Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB16329-50EA 22520 CZK
ABCAAB16329-50
Anti-TNFRSF10B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR