Tisk…

Anti-STAT1 alpha Rabbit polyclonal antibody

ABCAAB16353-50EA 15650 CZK
ABCAAB16353-50
Anti-STAT1 alpha Rabbit polyclonal antibody
Type: Primary
Antigen: STAT1 alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR