Tisk…

Anti-CAST Chicken polyclonal antibody

ABCAAB16423-50EA 16280 CZK
ABCAAB16423-50
Anti-CAST Chicken polyclonal antibody
Type: Primary
Antigen: CAST (calpastatin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR