Tisk…

Anti-POMP Chicken Polyclonal Antibody

Dodavatel: Abcam
ABCAAB16424-25EA 21140 CZK
ABCAAB16424-25
Anti-POMP Chicken Polyclonal Antibody
Protilátky
Type: Primary
Antigen: POMP (proteasome maturation protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR