Tisk…

Anti-PPP2R5C Sheep Polyclonal Antibody

Dodavatel: Abcam
ABCAAB16449-100EA 20130 CZK
ABCAAB16449-100
Anti-PPP2R5C Sheep Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PPP2R5C (protein phosphatase 2, regulatory subunit B', gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR