Tisk…

Anti-MAPK1/3 Sheep Polyclonal Antibody

Dodavatel: Abcam
ABCAAB16573-125EA 21140 CZK
ABCAAB16573-125
Anti-MAPK1/3 Sheep Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR