Tisk…

Anti-Orexin Prepro Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB16584-50EA 21300 CZK
ABCAAB16584-50
Anti-Orexin Prepro Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Orexin Prepro
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR