Tisk…

Anti-MKI67 Rabbit Monoclonal Antibody [clone: SP6]

Dodavatel: Abcam
ABCAAB16667-500EA 26080 CZK
ABCAAB16667-500
Anti-MKI67 Rabbit Monoclonal Antibody [clone: SP6]
Protilátky
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: monoclonal
Clone: SP6
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR