Tisk…

Anti-GPX4 Mouse monoclonal antibody [clone: 1H11]

Dodavatel: Abcam
ABCAAB16739-100EA 19800 CZK
ABCAAB16739-100
Anti-GPX4 Mouse monoclonal antibody [clone: 1H11]
Type: Primary
Antigen: GPX4 (glutathione peroxidase 4)
Clonality: monoclonal
Clone: 1H11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR