Tisk…

Anti-PRDX1 Mouse monoclonal antibody [clone: 9D2]

ABCAAB16747-100EA 17160 CZK
ABCAAB16747-100
Anti-PRDX1 Mouse monoclonal antibody [clone: 9D2]
Type: Primary
Antigen: PRDX1 (peroxiredoxin 1)
Clonality: monoclonal
Clone: 9D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR