Tisk…

Anti-TXNRD2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB16841-100EA 23080 CZK
ABCAAB16841-100
Anti-TXNRD2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TXNRD2 (thioredoxin reductase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR