Tisk…

Anti-RPA32/RPA2 Mouse monoclonal antibody [clone: RPA34-19]

ABCAAB16855-100EA 17250 CZK
ABCAAB16855-100
Anti-RPA32/RPA2 Mouse monoclonal antibody [clone: RPA34-19]
Type: Primary
Antigen: RPA32/RPA2
Clonality: monoclonal
Clone: RPA34-19
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR