Tisk…

Anti-PDGFRB Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB16861-100EA 22340 CZK
ABCAAB16861-100
Anti-PDGFRB Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR