Tisk…

Anti-TLR9 Mouse Monoclonal Antibody [clone: 5G5]

Dodavatel: Abcam
ABCAAB16892-25EA 21140 CZK
ABCAAB16892-25
Anti-TLR9 Mouse Monoclonal Antibody [clone: 5G5]
Protilátky
Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: monoclonal
Clone: 5G5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR