Tisk…

Anti-FOXO3 Goat polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB17026-100EA 23320 CZK
ABCAAB17026-100
Anti-FOXO3 Goat polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: FOXO3 (forkhead box O3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR