Tisk…

Anti-liver FABP Mouse monoclonal antibody [clone: K5A6]

ABCAAB17159-500EA 16280 CZK
ABCAAB17159-500
Anti-liver FABP Mouse monoclonal antibody [clone: K5A6]
Type: Primary
Antigen: liver FABP
Clonality: monoclonal
Clone: K5A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR