Tisk…

Anti-HPV16 E2 Mouse Monoclonal Antibody [clone: TVG 261]

Dodavatel: Abcam
ABCAAB17185-50EA 23230 CZK
ABCAAB17185-50
Anti-HPV16 E2 Mouse Monoclonal Antibody [clone: TVG 261]
Protilátky
Type: Primary
Antigen: HPV16 E2
Clonality: monoclonal
Clone: TVG 261
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR