Tisk…

Anti-F9 Mouse monoclonal antibody unconjugated [clone: 1C2]

Dodavatel: Abcam
ABCAAB17196-100EA 22520 CZK
ABCAAB17196-100
Anti-F9 Mouse monoclonal antibody unconjugated [clone: 1C2]
Protilátky
Type: Primary
Antigen: F9 (coagulation factor IX)
Clonality: monoclonal
Clone: 1C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR