Tisk…

Anti-TRIM5 gamma Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB17203-100EA 20380 CZK
ABCAAB17203-100
Anti-TRIM5 gamma Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRIM5 gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR