Tisk…
Anti-Trypsin Mouse Monoclonal Antibody [clone: 2C4]
Dodavatel: Abcam
ABCAAB17263-100EA 27330 CZK
ABCAAB17263-100
Anti-Trypsin Mouse Monoclonal Antibody [clone: 2C4]
Protilátky
Type: Primary
Antigen: Trypsin
Clonality: monoclonal
Clone: 2C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human