Tisk…

Anti-Ovalbumin Mouse Monoclonal Antibody [clone: 2D11]

Dodavatel: Abcam
ABCAAB17290-100EA 22490 CZK
ABCAAB17290-100
Anti-Ovalbumin Mouse Monoclonal Antibody [clone: 2D11]
Protilátky
Type: Primary
Antigen: Ovalbumin
Clonality: monoclonal
Clone: 2D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR