Tisk…

Anti-APP Goat polyclonal antibody

ABCAAB17325-100EA 16280 CZK
ABCAAB17325-100
Anti-APP Goat polyclonal antibody
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR