Tisk…
Anti-HIST2H2AC Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB17353-100EA 26940 CZK
ABCAAB17353-100
Anti-HIST2H2AC Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HIST2H2AC (histone cluster 2, H2ac)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat