Tisk…

Anti-MYC Mouse Monoclonal Antibody [clone: 8]

Dodavatel: Abcam
ABCAAB17355-100EA 27670 CZK
ABCAAB17355-100
Anti-MYC Mouse Monoclonal Antibody [clone: 8]
Protilátky
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR