Tisk…

Anti-BCL6 Mouse monoclonal antibody [clone: P1F6]

ABCAAB17402-6EA 16280 CZK
ABCAAB17402-6
Anti-BCL6 Mouse monoclonal antibody [clone: P1F6]
Type: Primary
Antigen: BCL6 (B-cell CLL/lymphoma 6)
Clonality: monoclonal
Clone: P1F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR