Tisk…

Anti-IL20 Goat polyclonal antibody

ABCAAB17426-50EA 16160 CZK
ABCAAB17426-50
Anti-IL20 Goat polyclonal antibody
Type: Primary
Antigen: IL20 (interleukin 20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR