Tisk…

Anti-C3d Mouse monoclonal antibody unconjugated [clone: 7C10]

Dodavatel: Abcam
ABCAAB17453-100EA 23110 CZK
ABCAAB17453-100
Anti-C3d Mouse monoclonal antibody unconjugated [clone: 7C10]
Protilátky
Type: Primary
Antigen: C3d
Clonality: monoclonal
Clone: 7C10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR