Tisk…

Anti-RETN Goat polyclonal antibody Biotin

ABCAAB17592-50EA 16280 CZK
ABCAAB17592-50
Anti-RETN Goat polyclonal antibody Biotin
Type: Primary
Antigen: RETN (resistin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR