Tisk…

Anti-NAP1L4 Goat polyclonal antibody

ABCAAB17596-50EA 16280 CZK
ABCAAB17596-50
Anti-NAP1L4 Goat polyclonal antibody
Type: Primary
Antigen: NAP1L4 (nucleosome assembly protein 1-like 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR