Tisk…

Anti-MCM2 Mouse monoclonal antibody [clone: SPM196]

ABCAAB17788-7EA 16480 CZK
ABCAAB17788-7
Anti-MCM2 Mouse monoclonal antibody [clone: SPM196]
Type: Primary
Antigen: MCM2 (minichromosome maintenance complex component 2)
Clonality: monoclonal
Clone: SPM196
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR