Tisk…

Anti-CASP3 Mouse monoclonal antibody [clone: 3G2]

ABCAAB17815-50EA 16280 CZK
ABCAAB17815-50
Anti-CASP3 Mouse monoclonal antibody [clone: 3G2]
Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 3G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR