Tisk…

Anti-MYB Mouse monoclonal antibody [clone: SPM175]

ABCAAB17851-500EA 16480 CZK
ABCAAB17851-500
Anti-MYB Mouse monoclonal antibody [clone: SPM175]
Type: Primary
Antigen: MYB (v-myb avian myeloblastosis viral oncogene homolog)
Clonality: monoclonal
Clone: SPM175
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR