Tisk…
Anti-Factor B Mouse Monoclonal Antibody [clone: 6G11]
Dodavatel: Abcam
ABCAAB17927-100EA 26940 CZK
ABCAAB17927-100
Anti-Factor B Mouse Monoclonal Antibody [clone: 6G11]
Protilátky
Type: Primary
Antigen: Factor B
Clonality: monoclonal
Clone: 6G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human