Tisk…

Anti-Sodium Iodide Symporter Mouse monoclonal antibody [clone: SPM186]

ABCAAB17950EA 16480 CZK
ABCAAB17950
Anti-Sodium Iodide Symporter Mouse monoclonal antibody [clone: SPM186]
Type: Primary
Antigen: Sodium Iodide Symporter
Clonality: monoclonal
Clone: SPM186
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR