Tisk…

Anti-ADM Mouse monoclonal antibody [clone: HTA91a/G2]

ABCAAB18093-50EA 17160 CZK
ABCAAB18093-50
Anti-ADM Mouse monoclonal antibody [clone: HTA91a/G2]
Type: Primary
Antigen: ADM (adrenomedullin)
Clonality: monoclonal
Clone: HTA91a/G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR