Tisk…

Anti-RPSA Mouse monoclonal antibody [clone: SPM278]

ABCAAB18099-7EA 16480 CZK
ABCAAB18099-7
Anti-RPSA Mouse monoclonal antibody [clone: SPM278]
Type: Primary
Antigen: RPSA (ribosomal protein SA)
Clonality: monoclonal
Clone: SPM278
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR