Tisk…

Anti-DNM1 Sheep polyclonal antibody

ABCAAB18101-50EA 16080 CZK
ABCAAB18101-50
Anti-DNM1 Sheep polyclonal antibody
Type: Primary
Antigen: DNM1 (dynamin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR