Tisk…

Anti-FOS Rabbit polyclonal antibody

ABCAAB18119-50EA 17160 CZK
ABCAAB18119-50
Anti-FOS Rabbit polyclonal antibody
Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR