Tisk…
Anti-TUBB3 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB18207-100EA 29480 CZK
ABCAAB18207-100
Anti-TUBB3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TUBB3 (tubulin, beta 3 class III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat