Tisk…

Anti-CD19 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LT19]

Dodavatel: Abcam
ABCAAB18224-50EA 22340 CZK
ABCAAB18224-50
Anti-CD19 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LT19]
Protilátky
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: LT19
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR